Search
Direct Reverse Transcription: From Cells-to-cDNA |
Direct reverse transcription kits are designed to synthesize cDNA directly from cell lysates without isolating RNA. Lysis and reverse transcription are performed in the same tube. The resulting first-strand cDNA is ready to use in downstream applications including PCR, qPCR, and NGS.
|
| |
| Intended use | First-strand cDNA synthesis directly from mammalian cell lysates | Template switching reverse transcription and global preamplification of full-length cDNA |
| RNA purification required | No | No |
| Starting material | 1–10,000 cells | 1–1,000 cells 2 pg–10 ng total RNA |
| Final reaction product | cDNA | PCR amplified cDNA |
| Workflow | ![]() | ![]() |
| Reagents included | Lysis → RT | Lysis → RT → PreAmp |
| RT primer | Random + oligo(dT) | Oligo(dT)-based |
| DNA removal | Yes (DNase digestion) | Yes (mRNA enrichment) |
| Number of targets | Limited | Full transcriptome |
| Sensitivity | + | +++ |
| Yield | + | +++ |
| Applications | RT-PCR, RT-qPCR, cloning | Global preamplification, RNA-seq, RT-qPCR |
| Order now | Order now |
The cDNA synthesis master mix from the SuperScript IV Single Cell/Low-Input cDNA PreAmp kit is now available as a standalone SuperScript IV Template Switching RT Master Mix.
This master mix is designed for high template switching efficiency in cDNA synthesis reactions and contains SuperScript IV Reverse Transcriptase, RNase Inhibitor, and all other reverse transcription reaction components in a convenient premixed format. Only RNA template, RT primer, and template switching oligo (TSO) of choice need to be added. The master mix is compatible with a wide range of total RNA (from 2 pg to 4 µg) and is supplied with lysis buffer to enable direct cDNA synthesis from intact mammalian cells (1–1,000 cells per sample).
In conjunction with SuperScript IV Template Switching RT Master Mix, we offer Template Switching Oligo and Capturing Oligo (dT) Primer, both containing adaptor sequences complementary to the Preamplification Primer.
In the template switching mechanism, a reverse transcriptase enzyme binds to the RNA template and RT primer duplex and begins synthesizing a complementary DNA strand. When the reverse transcriptase reaches the 5’ end of the RNA template, terminal deoxynucleotidyl transferase (TdT) activity adds 1–3 additional nucleotides to the cDNA end, enabling binding of the template switching oligo and incorporation of known sequence at the 3’ end of the resulting cDNA (Figure 1). This cDNA can be used for amplification by PCR, gene expression analysis by real-time PCR, or as a template for 5’RACE, RNA-seq, or second strand cDNA synthesis.
The SuperScript IV Single Cell/Low-Input cDNA PreAmp Kit workflow takes significantly less time when compared to workflows with competitor products (Figure 2).
Figure 2. Workflow comparisons with competitors. Workflows and timelines for the SuperScript IV Single Cell/Low-Input cDNA PreAmp Kit in comparison to the Smart-seq3 Protocol V.3 workflow, NEBNext™ kit workflow, and Takara SMART-Seq v4™.
Experimental data support that SuperScript IV Single Cell/Low-Input cDNA PreAmp Kit generates high cDNA yields from low-input RNA or single-cell RNA. cDNA yields from low-input (2 pg) Universal Human Reference RNA (UHRR) were almost twice the yield from NEBNext™ Single Cell/Low Input cDNA Synthesis and Amplification Module (Figure 3).
Figure 3. Comparison of cDNA yields from SuperScript IV PreAmp kit with NEBNext™ preamplification kit. SuperScript IV PreAmp kit produces excellent cDNA yield, compared to the NEBNext™ Single Cell/Low Input cDNA Synthesis and Amplification Module. Error bars represent standard deviation among at least five replicates.
SuperScript IV PreAmp kit uniformly preamplifies transcripts for reliable RNA sequencing. Libraries prepared from cDNA generated with the SuperScript IV PreAmp kit resulted in low bias across most of the normalized transcript distance (Figure 4).
Figure 4. Normalized transcript coverage across several different input types. Single-cell RNA was reverse transcribed and the obtained cDNA was preamplified using the recommended number of PCR cycles. The Invitrogen Collibri ES DNA Library Prep Kit for Illumina™ Systems with UD indexes was used to prepare libraries for sequencing using the MiSeq™ Sequencing system.
The SuperScript IV Single Cell/Low-Input cDNA PreAmp Kit workflow takes significantly less time when compared to workflows with competitor products (Figure 2).
Figure 2. Workflow comparisons with competitors. Workflows and timelines for the SuperScript IV Single Cell/Low-Input cDNA PreAmp Kit in comparison to the Smart-seq3 Protocol V.3 workflow, NEBNext™ kit workflow, and Takara SMART-Seq v4™.
Experimental data support that SuperScript IV Single Cell/Low-Input cDNA PreAmp Kit generates high cDNA yields from low-input RNA or single-cell RNA. cDNA yields from low-input (2 pg) Universal Human Reference RNA (UHRR) were almost twice the yield from NEBNext™ Single Cell/Low Input cDNA Synthesis and Amplification Module (Figure 3).
Figure 3. Comparison of cDNA yields from SuperScript IV PreAmp kit with NEBNext™ preamplification kit. SuperScript IV PreAmp kit produces excellent cDNA yield, compared to the NEBNext™ Single Cell/Low Input cDNA Synthesis and Amplification Module. Error bars represent standard deviation among at least five replicates.
SuperScript IV PreAmp kit uniformly preamplifies transcripts for reliable RNA sequencing. Libraries prepared from cDNA generated with the SuperScript IV PreAmp kit resulted in low bias across most of the normalized transcript distance (Figure 4).
Figure 4. Normalized transcript coverage across several different input types. Single-cell RNA was reverse transcribed and the obtained cDNA was preamplified using the recommended number of PCR cycles. The Invitrogen Collibri ES DNA Library Prep Kit for Illumina™ Systems with UD indexes was used to prepare libraries for sequencing using the MiSeq™ Sequencing system.
SuperScript IV CellsDirect cDNA synthesis kit requires a simplified cDNA synthesis workflow that eliminates the need to purify RNA (Figure 5). With this approach, fewer pipetting steps are required, which helps prevent RNA loss and contamination, saving time and effort. In addition, SuperScript IV CellsDirect cDNA Synthesis Kit delivers exceptional sensitivity, yield, and linearity.
Figure 5. Comparison workflow versus traditional methods. The protocol used for SuperScript IV CellsDirect cDNA synthesis kit goes from cells-to-cDNA in less than half the time it takes to produce cDNA using traditional methods; the time savings are associated with RNA purification.
The SuperScript IV CellsDirect cDNA synthesis kit is sensitive across a range of cell types and sample sizes. It consistently outperforms kits from competitors by producing higher yields for abundant (ACTB), moderately abundant (PGK1), and low abundance (BCL2) transcripts (Figure 6).
Figure 6.Comparative cDNA yields among direct RT kits for a variety of cellular transcripts. Samples of three distinct mammalian cell types (Jurkat, K562, and iPSCs) were used for cDNA generation using the SuperScript IV CellsDirect kit and four other commercially available direct RT kits. qPCR was performed to evaluate cDNA yields from 44 different RNA transcripts, representing a wide range of cellular abundance. The SuperScript IV CellsDirect kit had the highest cDNA yields
The SuperScript IV CellsDirect kit demonstrated the strongest linear correlation across four mRNA targets (ACTB, BCL2, PGK1, PPIA) across serial dilution of HeLa S3 cells ranging from 1 to 10,000 cells (Figure 7).
SuperScript IV CellsDirect cDNA synthesis kit requires a simplified cDNA synthesis workflow that eliminates the need to purify RNA (Figure 5). With this approach, fewer pipetting steps are required, which helps prevent RNA loss and contamination, saving time and effort. In addition, SuperScript IV CellsDirect cDNA Synthesis Kit delivers exceptional sensitivity, yield, and linearity.
Figure 5. Comparison workflow versus traditional methods. The protocol used for SuperScript IV CellsDirect cDNA synthesis kit goes from cells-to-cDNA in less than half the time it takes to produce cDNA using traditional methods; the time savings are associated with RNA purification.
The SuperScript IV CellsDirect cDNA synthesis kit is sensitive across a range of cell types and sample sizes. It consistently outperforms kits from competitors by producing higher yields for abundant (ACTB), moderately abundant (PGK1), and low abundance (BCL2) transcripts (Figure 6).
Figure 6.Comparative cDNA yields among direct RT kits for a variety of cellular transcripts. Samples of three distinct mammalian cell types (Jurkat, K562, and iPSCs) were used for cDNA generation using the SuperScript IV CellsDirect kit and four other commercially available direct RT kits. qPCR was performed to evaluate cDNA yields from 44 different RNA transcripts, representing a wide range of cellular abundance. The SuperScript IV CellsDirect kit had the highest cDNA yields
The SuperScript IV CellsDirect kit demonstrated the strongest linear correlation across four mRNA targets (ACTB, BCL2, PGK1, PPIA) across serial dilution of HeLa S3 cells ranging from 1 to 10,000 cells (Figure 7).
Not sure if direct or template switching reverse transcription is right for you?
Reverse transcriptase selection tool
Find the right products to fit your cDNA synthesis needs.
Reverse transcriptase webinars
Stay up-to-date on current industry trends in cDNA synthesis applications with on-demand webinars.
Reverse transcriptase literature
Access white papers and technical and application notes for insights on cutting edge reverse transcription technology.
Reverse transcriptase videos
Watch instructional videos on troubleshooting tips, how-to videos, and explainers related to cDNA synthesis.
Reverse transcriptase education
Review educational resources on topics including reverse transcription basics, cDNA synthesis, enzyme selection, troubleshooting tips, and cDNA applications in molecular biology.
PCR and cDNA Synthesis Support Center
Find support for every step of your cDNA synthesis experiments.
Tm calculator
Handy tool to calculate Tm (melting temperature) of primers and annealing temperatures.
Contact us
Get in touch with our technical application scientists for questions regarding reverse transcription enzymes, master mixes, and cDNA synthesis kits.
Mobile and desktop applications
Access content and helpful tools through SmartIST and Cloning Bench mobile apps.
OEM and custom solutions
Source reverse transcriptase and other molecular biology enzymes, PCR reagents, and plastics customized to your needs.
For Research Use Only. Not for use in diagnostic procedures.