SuperScript™ IV CellsDirect™ Lysis Reagents
Invitrogen™

SuperScript™ IV CellsDirect™ Lysis Reagents

The SuperScript IV CellsDirect Lysis Reagents kit includes lysis solution, stop solution, and lysis enhancer. These reagents are part of the SuperScript IV CellsDirect cDNA Synthesis Kit and are made available separately here for use when processing a large number of samples.
Have Questions?
카탈로그 번호수량
11750550500 reactions
카탈로그 번호 11750550
제품 가격(KRW)
2,559,000
온라인 행사
Ends: 31-Dec-2026
3,010,000
할인액 451,000 (15%)
Each
수량:
500 reactions
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
2,559,000
온라인 행사
Ends: 31-Dec-2026
3,010,000
할인액 451,000 (15%)
Each

The SuperScript IV CellsDirect Lysis Reagents kit includes lysis solution, stop solution, and lysis enhancer. These reagents are part of the SuperScript IV CellsDirect cDNA Synthesis Kit and are made available separately here for use when processing a large number of samples. See the SuperScript IV CellsDirect cDNA Synthesis Kit manual for protocol recommendations.

SuperScript IV CellsDirect Lysis Reagents are optimized for synthesizing first-strand cDNA directly from a mammalian cell lysate without first isolating RNA.

Features of SuperScript IV CellsDirect Lysis Reagents include:

  • Ready-to-use pre-mixed cell lysis solution compatible with a variety of cell-types
  • Optimized performance for 1–10,000 cells per sample; results equivalent to those from purified RNA

Note:
This procedure is optimized for a maximum of 10,000 cells per reaction. If needed, volumes can be adjusted for cell numbers greater than 10,000. Using too many cells per lysis reaction can result in incomplete lysis and/or inhibition of RT-PCR.

사양
용도SuperScript IV CellsDirect cDNA Synthesis Kit
형식Kit
반응 수500 reactions
수량500 reactions
반응 형식Master Mix
시약 유형Reverse Transcription
역전사 효소SuperScript IV
배송 조건Dry Ice
시작 물질Cells
기술Reverse Transcription
Unit SizeEach
구성 및 보관

• SuperScript IV CellsDirect Lysis Solution (9 mL)
• SuperScript IV CellsDirect Stop Solution (1.25 mL)
• Lysis Enhancer (250 μL)

Store at –20°C.