Search
Primary XPS region: La3d
Overlapping regions: Transition metal Augers, La MNN
Binding energies of common chemical states:
| Chemical state | Binding energy La3d5/2 |
|---|---|
| La2O3 | 834.5 eV |
| La(OH)3 | 834.7 eV |
| La2(CO3)3 | 835 eV |
Symbol: La
Date of discovery: 1839
Name origin: Greek lanthanein
Appearance: white
Discoverer: Carl Gustaf Mosander
Obtained from: monazite, bastnasite
Melting point: 1193 K
Boiling point: 3743 K
Density[kg/m3]: 6146
Molar volume: 22.39 × 10-6 m3/mol
Protons/Electrons: 57
Neutrons: 82
Shell structure: 2,8,18,18,9,2
Electron configuration: [Xe]5d16s2
Oxidation state: 3
Crystal structure: hexagonal
This element is so soft that it can be cut by a knife. Lanthanum has an important role in the motion picture industry because it is used in studio lighting and projection. It is also used to make infrared absorbing glass and is used in the lenses of cameras and telescopes. Adding small amounts of lanthanum to steel improves steel’s malleability and adding it to molybdenum decreases molybdenum’s hardness and sensitivity to temperature variation. While lanthanum has no known biological role, lanthanum chloride has anticoagulant properties. Injecting lanthanum solutions in animals causes glycohemia, low blood pressure, degeneration of the spleen, and hepatic alterations.
Products
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.