Treat
Coeliac 
Left untreated, coeliac disease may result in serious long-term health complications. The only effective treatment for coeliac disease is life-long adherence to a gluten-free diet,1 which can reverse villous atrophy and reduce associated morbidities.2
However, a gluten-free diet should only be initiated once all necessary serologic tests for coeliac disease have been finished.3
My patient has coeliac disease, confirmed by a gastroenterologist. What’s next?
There are several ways in which you can manage and support your patient at the time of their diagnosis:3,4
How should coeliac disease be monitored?
Patients are more likely to adhere to a gluten-free diet if they are regularly followed up in a specialist coeliac clinic, with input from a dietitian and a gastroenterologist.3
In the first year following a diagnosis follow-up should be frequent to increase the likelihood of dietary adherence, provide psychological support, and help the patient adapt to living with coeliac disease. Once the disease is stable, annual follow-up may be initiated.3
At each appointment, it’s recommended to check serum tissue transglutaminase (tTG) IgA levels.3
  • It’s common practice for tTG IgA to be tested 2-3 times per year until levels normalise, then once a year to monitor disease activity.5
  • A decline in tTG IgA titers is an indication of good dietary adherence.3
It’s also important to monitor:3
  • Symptoms and coping skills
  • The gluten-free diet plan
  • Nutritional status, height, and weight
  • Blood markers, as clinically indicated (e.g., to look for associated autoimmune conditions)
IgA: immunoglobulin A 
Get news on the topic of your choice, right in your inbox.
1. Gujral N, Freeman H J, Thomson A B. Celiac disease: prevalence, diagnosis, pathogenesis and treatment. World J Gastroenterol 2012;18(42):6036-6059
2. Ciacci C, Ciclitira P et al. The gluten-free diet and its current application in coeliac disease and dermatitis herpetiformis. United European Gastroenterol J 2015;3(2):121-135
3. Al-Toma A, Volta U et al. European Society for the Study of Coeliac Disease (ESsCD) guideline for coeliac disease and other gluten-related disorders. United European Gastroenterol J 2019;7(5):583-613
4. Rubio-Tapia A, Hill I D et al. ACG clinical guidelines: diagnosis and management of celiac disease. Am J Gastroenterol 2013;108(5):656-676;quiz 677
5. Pinto-Sanchez M I, Bai J C. Toward new paradigms in the follow up of adult patients with celiac disease on a gluten-free diet. Front Nutr 2019;6:153