GC Autosamplers
A step ahead in automated sampling for GC analysis
If you need a simple liquid or headspace GC autosampler, or one that can do it all (liquid, headspace, solid phase microextraction (SPME) injections, and automated sample preparation), our autosamplers are a step ahead of the norm in performing liquid, solid, and gas sample handling and injection into GC and GC-MS with precision and reliability.
Browse our complete portfolio below, or let one of our experts help you tailor a selection specific to your application and workflow needs.
Contact us
Featured GC autosamplers
Process more samples quickly, while improving the consistency and quality of your GC data. This rugged, easy-to-use GC liquid autosampler is available as the 8-position AI 1610 autoinjector and the 155-sample capacity AS 1610 autosampler. Both are ideal in QA/QC and high-throughput environments. For extended productivity, the system can operate two injectors simultaneously via its Gemini configuration.
Automate liquid injections completely and reliably, using this X-Y-Z robotic autosampler. The TriPlus 100 LS makes the most of liquid injection with its high flexibility, which ranges from nano to large volume injection, and includes fully customizable injection parameters.
Boost lab productivity with advanced sample handling, automated liquid sampling, static headspace, dynamic headspace through In-Tube Extraction (ITEX), and SPME sampling and injection. Easily create and test sample preparation methods with the Thermo Scientific TriPlus RSH Sampling Workflow Editor Software. When connected to the TriPlus RSH SMART Autosampler, this software enables quick programming of common sample preparation steps, with automation of daily sample handling operations as well as more complex sample preparation procedures. The software also enables visual programming without coding due to its intuitive drag-and-drop user interface.
Streamline Volatile Organic Compounds headspace analysis in water and soil samples with a dedicated TriPlus RSH SMART platform in a dual-head configuration. To preserve sample integrity and ensure the best repeatability in compliance with regulatory requirements, the automated workflow includes the dilution of calibration standards and the addition of IS/surrogates just before incubation.
Fully controlled in Thermo Scientific Chromeleon CDS, this automated workflow removes the variability of manual sample handling, providing valuable labor-time saving and highly reliable data.
Automate and accelerate your analyses of organic volatiles. The TriPlus 500 GC Headspace Autosampler helps increase sample turnaround and lower overall cost per analysis. Based on Valve-and-Loop technology, this autosampler also ensures the highest data quality and full compliance with regulatory statutes.
Featured video
The TriPlus 500 GC Headspace Autosampler: Reliable unattended operations maximizes time and expanded vial capability optimizes throughput.
Our Products have not been tested or validated for such applications...
The LC-MS are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our Products have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in inaccurate results and, more seriously, health and safety risks. They are designated for Research Use Only Applications.
CMD SchemaApp code