Protein detection and analysis—western blotting
Our all-encompassing range of solutions for western blotting analysis is meticulously designed and tested to meet the highest standards and minimize time and effort. Establish a robust foundation for your western blotting experiments.
Innovative western blotting solutions
Boost your productivity with western blotting tools designed for efficient high-quality data generation. 
iGlow Gel Documentation System
Simple, efficient imaging and documentation for routine protein and nucleic acid gels when western blot is not required.
  • Simple—capture high-quality gel images with one touch
  • Flexible—compatible with various gel sizes of protein and nucleic acid gels, including Invitrogen E-Gel DNA gels and precast mini and midi protein gels, and pour-your-own (PYO) gels
  • Accessible—beginner friendly and user installable, suitable for everyday gel imaging needs in individual labs
iGlow Gel Documentation System
Simple, efficient imaging and documentation for routine protein and nucleic acid gels when western blot is not required.
  • Simple—capture high-quality gel images with one touch
  • Flexible—compatible with various gel sizes of protein and nucleic acid gels, including Invitrogen E-Gel DNA gels and precast mini and midi protein gels, and pour-your-own (PYO) gels
  • Accessible—beginner friendly and user installable, suitable for everyday gel imaging needs in individual labs
Product combinations for western blot success
Help improve the success rate of your western blotting experiments by starting with carefully curated product combinations. Designed to work easily together to help improve lab efficiency and support your research.
Western blot of high molecular weight proteins
Begin with excellent protein separation and efficient protein transfer with NuPAGE Tris-Acetate Gels and iBlot 3 Western Blot Transfer System.
Application note: Optimizing western blot transfer for high and low molecular weight proteins
Run high performance protein gels in Bio-Rad’s Tank
Get sharp, straight bands no matter your protein molecular weight with Tetra Electrophoresis Cores and Invitrogen Precast Protein gels.
Easy total protein normalization
Streamline the quantitative western blotting workflow and help overcome the limitations of housekeeping protein-based normalization with iBright FL1500 Imaging System and No-Stain Protein Labeling Reagent.
Application note: Easy and accurate normalization of western blot data
Enhanced detection of low abundance proteins or dilute samples
Double your sample loading volume and help improve the sensitivity of chemiluminescent detection with WedgeWell format protein gels and SuperSignal West Atto Ultimate Sensitivity Substrate.
Everyday essentials for western blotting
Limit the variables in your experiments by using dependable products from one trusted source.
Speed, performance and convenience
Total protein normalization
Total protein normalization (TPN) for quantitative western blotting analysis offers several advantages over traditional housekeeping protein-based (HKP) normalization. One optimal protein labeling reagent for TPN is the No-Stain Protein Labeling Reagent. TPN using No-Stain Labeling Reagent exhibits good linearity of response within the concentration range typically investigated (1–80 µg of protein load). The wider dynamic range prevents the loss of valuable quantitative data due to signal saturation, a common occurrence with HKP normalization. TPN using No-Stain Labeling Reagent simplifies experimental design, helping save time and resources. Unlike HKP normalization, there is no need to verify that the expression of housekeeping proteins remains unaffected by the treatment. No-Stain Labeling Reagent is particularly well-suited for TPN due to its rapid labeling protocol, and compatibility with downstream transfer and immunoblotting. It enables sensitive and stable signal detection down to 20 ng of proteins, allowing for accurate quantitation of protein expression levels.
Learn more about TPN with No-Stain Labeling Reagent
Multiplexed fluorescent western blotting
Multiplexed western blotting is a powerful technique that offers several advantages over traditional western blotting. Its ability to simultaneously detect and quantify multiple proteins within a single sample helps increase efficiency, improves accuracy, reduces costs, and enhances data quality. It is advantageous when investigating complex biological pathways involving proteins with diverse molecular weights. The simultaneous detection of multiple protein targets in one experiment reduces experimental variability and is an excellent choice when the sample volume is limited or when analyzing large sample sets. Fluorescent western blotting requires careful planning and experiment design. Selecting the right products from the start reduces optimization and troubleshooting time and helps ensure the success of your experiment.
Design your next multiplex western blotting experiment
Protein electrophoresis and western blotting handbooks
The protein gel electrophoresis and western blotting handbooks are comprehensive, easy-to-use guides covering every procedural step from sample prep to imaging and data analysis.
Handbook covers
Western blot learning and troubleshooting
Whether you are new to western blotting, or an experienced researcher wanting to confirm your knowledge, access our educational tips and resource materials to help with every aspect of western blot analysis.
Access western blotting learning center
video icon
Videos
Discover the new ways to do western blotting with our how-to videos.
clipboard with checkmark icon
Protocols
Find recipes for buffers, stock solutions, and general western blot protocols.
book catalog internal serach icon
Foundation Library
Tech tips, application notes, handbooks and more.
gears troubleshooting icon
Troubleshooting
Overcome western blot difficulties with our troubleshooting guidance.
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.