- Correct orientation of the binding domain by directed biotinylation of the ligand.
- Minimal conformational changes of the immobilised target.
- Increased surface area allows for panning of saturated phage solutions.
- A higher percentage of positive binders and fewer rounds of panning.
- Magnetic separation allows for automation high throughput panning.
Phage display library screening is a valuable tool for protein-protein interaction mapping. Dynabeads® Streptavidin have also been used to generate antigen-specific monoclonal antibodies in vitro. Ligands are immobilized on the beads, and high-affinity binders selected and matured by using a hypermutating B-cell line with only four rounds of sorting (5).
Dynabeads® are widely used in automated devices for panning for drug candidates, and a number of companies worldwide are benefiting from the unique properties of the Dynabeads® Streptavidin.