Silver • Transition Metal
Primary XPS region: Ag3d
Overlapping regions: N/A
Binding energies of common chemical states:
Chemical stateBinding energy Ag3d5/2
Bromyrite (AgBr)~367.5 eV
Ag metal368.2 eV
Experimental information
  • This element has a high affinity for sulfur.
  • Silver compounds are typically X-ray sensitive.
Interpretation of XPS spectra
  • Ag3d region has well-separated spin-orbit components (Δmetal=6.0eV).
    • Peaks have asymmetric peak shape for metal.
    • Loss features are observed to higher binding energy side of each spin-orbit component for Ag metal.
  • Small binding energy shifts for compounds, such as oxides.
    • Ag3d peaks broaden relative to metal peaks.
element-silver-xpsspectra-600x400-v3
About this element
Element Crystal fcc
Symbol: Ag
Date of discovery: ancient times
Name origin: Anglo-Saxon siolfur
Appearance: silver
Discoverer: unknown
Obtained from: ores
Melting point: 1234.93 K
Boiling point: 2435 K
Density[kg/m3]: 10490
Molar volume: 10.27 × 10-6 m3/mol
Protons/Electrons: 47
Neutrons: 61
Shell structure: 2,8,18,18,1
Electron configuration: [Kr]4d105s1
Oxidation state: 1
Crystal structure: cubic face centered
Silver’s main use is as a precious metal. Sterling silver, over 90% silver, is used in flatware and jewelry. Silver’s malleability and ductility, physical appearance, and value make it a common material in coinage. Of all metals, silver has the highest electrical and thermal conductivity, but because of silver’s high cost, copper is commonly used in electrical applications. United States coins were minted of silver until after 1964. Silver’s halide salts are widely used in photography. The South American country, Argentina, is known as a source of silver and takes its name from the Latin word for silver, argentum.
element_silver
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.