Thermo Fisher Scientific offers an extensive line of SYBR® Green products to meet all of your Real-Time PCR needs.
Complementary Products
Primer Design
cDNA Synthesis
Our design tools are used for designing SYBR® primers and are seamlessly connected to our online ordering system
Our design tools are used for designing SYBR® primers and are seamlessly connected to our online ordering system.
Get the SuperScript® VILO™ cDNA Synthesis Kit.
Learn moreShop now
Alternative Products
Fast Master Mix
One Step Kits
This Master Mix allows you to complete 35 min runs.
With one-step RT-PCR kits you can perform reverse transcription and PCR all in one reaction tube, minimizing your hands on time and eliminating the need for reagent optimization.
Fast SYBR® Green Master Mix ›Power SYBR® Green RNA-to-CT™ 1-Step Kits ›
RT-PCR Directly From Cells
Cells-to-CT Kits take you from cultured cells directly to Real-Time PCR results, eliminating the need for laborious and time consuming RNA purification. Fast SYBR® Green Cells-to-CT Kits are optimized for Fast instruments, while the Power SYBR® Green Cells-to-CT Kits offer greater sensitivity for standard Real-Time PCR.
Power SYBR® Green Cells-to-CT™ Kits ›
Fast SYBR® Green Cells-to-CT™ Kits ›
For Research Use Only. Not for use in diagnostic procedures.