Search
The Thermo Scientific Hypulse Surface Analysis System supplements traditional ion-beam methods with femtosecond laser ablation (fs-LA) technology to offer outstanding X-ray photoelectron spectroscopy (XPS) depth profiling capabilities for thin films, coatings, and multi-layer structures. Fs-LA XPS depth profiling provides even more accurate, efficient, and versatile analysis from the surface to interfaces deep with the sample. This helps you optimize fabrication and the properties of technologically important materials and devices, develop new multi-functional materials, and gain a deeper understanding of material failures.
Fs-LA XPS delivers accurate, reliable surface and interface analysis without the risk of chemical damage to the sample. With this cutting-edge technique, you can quickly, efficiently, and precisely measure technologically important materials and devices to better understand their structure.
Designed for precision and accuracy, the Hypulse System allows you to explore the intricate layers of diverse materials while preserving their chemistry during the analysis. Whether you are in the electronics, metallurgy, or surface coatings industries, it provides fast, detailed, and accurate analysis.
Accurate and efficient chemical analysis is essential for materials science research labs, academic institutions, and industries such as semiconductor manufacturing, polymer engineering, coating development, and the development of new multifunctional materials. Traditional ion gun-based methods for X-ray photoelectron spectroscopy depth profiling can have issues with certain materials, which results in data that is not completely representative of the sample.
By combining monatomic ion beam etching, cluster ion beam etching, and fs-LA depth profiling, the Hypulse System can provide accurate results for more materials than was previously possible.
Fs-LA depth profiling can perform multiple profiles on a material in the time that one sample can be processed using ion beam profiling.
The Hypulse System can help you practically measure depth profiles that extend beyond 10 µm into the sample. Buried layers that would take significant time to discover using ion beam profiling can be accessed more easily through fs-laser ablation depth profiling.
Rather than choosing between cluster ions or monatomic ions, the Hypulse System’s laser settings can be tuned to the material, making it easier to profile complex composite samples.
The Hypulse System is built around a fully integrated 1,030 nm fs-laser system and high-quality optical components. The laser is focused on the analysis position and can be attenuated through Thermo Scientific Avantage Software. The instrument is a class 1 laser system, so no special laser requirements are necessary for the laboratory.
To complement the fs-LA system, the Thermo Scientific MAGCIS Dual Beam Ion Source for XPS Instruments is included as standard. Automatic source optimization and gas handling ensure excellent performance and experimental reproducibility.
The Hypulse System is a high-performing XPS instrument and includes additional analytical techniques such as REELS and ISS to help you get the most information from your samples.
The patented dual-beam flood source couples low-energy ion beams with very low-energy electrons (less than 1 eV) to prevent sample charging during analysis, which eliminates the need for charge referencing and makes analyzing data from insulating samples easy and reliable.
Expand your possibilities with available specialist sample holders for angle-resolved XPS, sample bias measurements, sample heating, or inert transfer from a glove box.
Instrument control, data processing, and reporting are all controlled by Avantage Software. The software includes Knowledge View, a comprehensive source of tutorials and reference material that guides you through analysis.
Correlate XPS data with SEM images through the correlative imaging and surface analysis (CISA) workflow using Thermo Scientific Maps Software.
| Analyzer type |
|
| X-ray source |
|
| X-ray spot size |
|
| Laser ablation system |
|
| Ion depth profiling |
|
| Additional included analytical methods |
|
| Maximum sample area |
|
| Maximum sample thickness |
|
Vacuum system |
|
| Optional accessories |