PrimeFlow
Detecting RNA Targets by Flow Cytometry
Invitrogen PrimeFlow RNA Assay expands the capability of flow cytometry to measure RNA. Combine PrimeFlow with immunolabeling of both cell-surface and intracellular proteins using fluorophore-conjugated antibodies. Researchers now can reveal the dynamics of RNA transcription together with protein expression patterns in single cell resolutions
Download brochure
Find PrimeFlow probes for your target 
Principle behind the PrimeFlow RNA assay
Featured PrimeFlow Assay applications
Featured PrimeFlow applications
Evaluate virus RNA in infected cells
Featured PrimeFlow applications
Detect microRNA
Featured PrimeFlow applications
Probe mRNA when no antibody exists to detect the protein
Not finding the application you're looking for? Let us know what you need
Ordering information
Follow the five steps below to begin your PrimeFlow RNA Assay
InstrumentSpecification
Flow cytometer
Lasers:
  • Blue (488 nm)
  • Yellow-green (561 nm)
  • Red (633 nm)
Filter sets:
  • FITC: 530/20
  • PE-eFluor-610: 610/20
  • APC: 660/20
  • APC-eFluor 780 (APC-Cyanine7): 780/60
IncubatorCapable of maintaining temperature at 40 ±1°C
Metal heat blockFor 1.5 mL microcentrifuge tube, placed inside the validated incubator
Temperature validation kitThe Invitrogen ViewRNA Temperature Validation Kit uses a calibrated thermometer to verify the incubator temperature
Optional: Is your target microRNA? This buffer helps ensure you get improved signal and better sensitivity for miRNA.
Optional: Will the assay be processed in a 96-well plate?
  • Four types of probe sets are currently available for RNA detection 
  • Select different types of probe set depending on the RNA expression level 
  • For multiplex analysis with immunolabeling of both cell-surface and intracellular proteins, find fluorophore-conjugated antibodies
Expression level of target RNAFluorochrome labelLaser excitation wavelength (nm)Bandpass filter (nm)Fluorescence channelProbe set type
LowAlexa Fluor 647633, 635660/20APC, Alexa Fluor 647, eFluor 660Type 1
LowAlexa Fluor 568561610/20PE-eFluor 610, Alexa Fluor 568, PE-Texas RedType 10
Medium-to-highAlexa Fluor 488488530/30FITC, Alexa Fluor 488Type 4
Medium-to-highAlexa Fluor 750633, 635780/60APC-Cy7, APC-eFluor 780, Alexa Fluor 750Type 6
Search for available (in stock) probe sets
Not finding what you're looking for? Go to Custom Branched DNA Probe Set Tool
  • Positive control—use positive control probe sets in every experiment to ensure proper assay performance: RPL13a for human leukocytes and β-actin (ACTB) for mouse tissues; for specific cell types and other recommended genes, refer to Appendix 5 in the product manual of the PrimeFlow RNA Assay Kit
  • Negative control—such as samples with the target specific probe omitted, or samples labeled with a probe against a target not expressed in the cells of interest (e.g., dapB, a bacterial gene); highly recommended
Search probes used for positive and negative controls in probe search tool in Step 4.
Webinars
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.
Hero banner dark gradient overlay