Search
The Thermo Scientific mySPIN mini centrifuges, our smallest benchtop platform, are compact and efficient laboratory devices designed to perform quick spin-downs of small sample volumes. They are suitable for a range of applications, from microfilter cell separation and HPLC protocols to PCR as well as clinical and diagnostic applications.
They are available in two speeds, 6,300 and 12,500 rpm and operate quietly with low vibration. Their tiny footprint makes such efficient use of space that they are excellent for a personal workstation. The mySPIN centrifuges can accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2 mL) for customized use.
| mySPIN 6 mini centrifuge | ||
|
|
|
6 x 2 mL maximum capacity and up to 6,000 rpm |
Includes mySPIN 6 mini centrifuge with accessories and tube rack, and is packaged in a handy transparent PC transport box for sample separations in the field. |
12 x 2 mL maximum capacity and up to 12,500 rpm |
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.