Fast5 bannner
May 2018
Volume V, Edition 2
The iQ Series of gas analyzers use MODBUS protocol which enables the user to perform the functions of reading the various concentrations and other measurement values, read the status of the digital outputs of the analyzer, to trigger or simulate the activation of a digital input to the instrument and to change operational settings in the instrument.
The MODBUS commands that are implemented are explained in detail in the iQ Communications Manual. iQ Series Communications Manual.
MODBUS register tables are easily found on thermofisher.com. To locate the most current list of MODBUS registers, go to the appropriate iQ instrument product page (see below) and scroll to the bottom of the page to the Documents section.
Thermo Scientific iQ Series MODBUS Register Tables:
Have an idea for a future Fast5 issue? Let us know ›