Search
Thermo Scientific polypropylene containers are specialized, food-grade containers that meet FDA food contact requirements. Our triple seal and twin valve design containers are airtight and leak resistant. The patented flip-top is integrally hinged to the container, allowing the user to open and close the container with one hand.
We design and build custom molds to your specific requirements and manufacture a variety of containers, which are available in light-repellent colors—including amber and black—that can be customized to meet the needs of your application.
We offer a wide range of economical flip-top containers and vials for a variety of applications.
We manufacture transport tubes and closures for a wide range of laboratory and biomaterial packaging applications.
Our veterinary and dairy industry polypropylene vials feature the same patented construction as our flip-top containers, and are ideal for milk sampling, mastitis testing, and other veterinary applications.
We manufacture single- and split-specimen collection and transport kits utilizing custom, high-speed automated assembly equipment. Our manufacturing process ensures the collection site that the product has never been opened or tampered with prior to collection.