Search
Primary XPS region: Mg1s
Overlapping regions: Cl LMM
Binding energies of common chemical states:
| Chemical state | Binding energy Mg1s |
|---|---|
| Mg metal | 1303 eV |
| Mg native oxide | 1304.5 eV |
| MgCO3 | 1305 eV |
Carbonate charge referenced to adventitious C1s peak at 284.8eV, oxide referenced to Mg metal.
Experimental information
Interpretation of XPS spectra
About this element
Symbol: Mg
Date of discovery: 1808
Name origin: Magnesia
Appearance: grayish
Discoverer: Sir Humphrey Davy
Obtained from: sea water
Melting point: 650 K
Boiling point: 1090 K
Density[kg/m3]: 1.738
Molar volume: 14.00 × 10-6 m3/mol
Protons/Electrons: 12
Neutrons: 12
Shell structure: 2,8,2
Electron configuration: [Ne]3s2
Oxidation state: 2
Crystal structure: hexagonal
Magnesium was present in the water of a certain farmer in Epsom, England, where it was found to heal rashes and other skin problems – thus the name Epsom salts. Magnesium is an abundant element and is essential to the cells of all known living organisms. As plants use magnesium in the production of chlorophylls, most green vegetables, such as spinach, are good sources of magnesium. Since magnesium is a lightweight metal, about 2/3 the weight of aluminum, automobile manufacturers use it to produce various car parts. A racing Corvette was built with magnesium body panels and for many years Porsche used a magnesium alloy in its engine blocks.
Products
Para garantir o desempenho ideal do sistema, fornecemos acesso a uma rede de especialistas em serviços de campo, suporte técnico e peças de reposição certificadas.