Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Potassium Chloride (KCl) | 75.0 | 200.0 | 2.6666667 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 200.0 | 1.4705882 |
| Sodium Chloride (NaCl) | 58.0 | 8000.0 | 137.93103 |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 2160.0 | 8.059702 |
| Other Components | |||
| EDTA | 416.0 | 457.6 | 1.1 |
| rProtease | confidential | n/a | |
25300 - 0.05% Trypsin - EDTA (1x), phenol red
Cat No:
25300054
,
25300062
,
25300096
,
25300104
,
25300120
25200 - 0.25% Trypsin-EDTA (1x), phenol red
Cat No:
25200056
,
25200072
,
25200114
15400 - 0.5% Trypsin-EDTA (10x), no phenol red
Cat No:
15400054
15050 - 0.25% Trypsin (1x), phenol red
Cat No:
15050057
,
15050065
15090 - 2.5% Trypsin (10x), no phenol red
Cat No:
15090046
25050 - 0.25% Trypsin (1x), phenol red
Cat No:
25050014
,
25050030
TrypLE™ Select (1X), no phenol red
Cat No:
12563011
,
12563029
TrypLE™ Express (1X), no phenol red
Cat No:
12604013
,
12604021
,
12604039
,
12604054
TrypLE™ Select CTS™
Cat No:
A1285901
TrypLE™ Express (1X), phenol red
Cat No:
12605010
,
12605028
,
12605036
,
12605093