MMDX Resources
Results Interpretation | Know the Risk of Injury and Rejection
Know the Risk of Injury and Rejection
MMDx-Heart and MMDx-Kidney systems use the level of expression of selected genes or gene set measurements to assess the probability of TCMR and ABMR as well as the extent of acute and chronic injury.
The report generated summarizes the key findings (e.g. inflammation, acute kidney injury and atrophy-fibrosis scores and different rejection classifier scores) as well as the overall molecular interpretation. The relationship of the new biopsy to its nearest molecular neighbors in the Reference Set estimates future graft survival. Many other classifiers are also run to establish details of ABMR. The net result of all these measurements is integrated by a novel method called archetype analysis, a form of cluster analysis.(1)
Objective and Actionable Data
The assessment reports provide you with the following information:
Rejection related
  • Risk of T cell-mediated rejection (TCMR)
  • Risk of Antibody-mediated rejection (ABMR)
  • Under-immunosuppression/non-adherence
  • Objective basis for therapeutic choices
Parenchymal injury & non-rejection disease:
  • Acute parenchymal injury
  • Irreversible injury (atrophy scarring)
  • Risk of progression to failure
Need more information?
Contact us about MMDx
References
  1. Reeve J, Böhmig GA, Eskandary F, Einecke G, Lefaucheur C, Loupy A, Halloran PF; MMDx-Kidney study group. Assessing rejection-related disease in kidney transplant biopsies based on archetypal analysis of molecular phenotypes. JCI Insight. 2017 Jun 15;2(12):e94197. doi: 10.1172/jci.insight.94197. PMID: 28614805; PMCID: PMC5470931.