Terasaki HLA Tissue Typing Trays
Designed to Accurately Determine HLA Cell Surface Antigens
Producing consistent, accurate HLA typing of Class I or Class II antigens, this selection of trays is available in a 72-well format. Each tray contains positive and negative controls and accommodates most complement dependent cytotoxicity (CDC) methods.
Terasaki HLA Tissue Typing Trays
Terasaki HLA Tissue Typing Trays are used to assign HLA Class I or Class II antigens accurately and quickly. The trays contain positive and negative controls and are designed for use with both staining techniques, dye exclusion and fluorescence. Each well of the tray contains 1 µl of specific antiserum and 5 µl of heavy mineral oil. In addition, all trays come with pre-screened rabbit complement.
  • Broad tray selection
  • 72 well format
  • Accurate typing results
  • Well-characterized antisera
  • Long-lasting lots
  • Free prescreened rabbit complement
Terasaki Dry HLA Tissue Typing Trays
The Class I and Class II Dry Tissue Typing Trays contain 1 µl of dried HLA monoclonal antibody in each well. As with the basic HLA trays, each Dry Tissue well contains a positive and negative control. Class II trays also include anti-B and anti-T lymphocyte controls.
Designed to withstand warmer shipping conditions and extended shipping periods, Terasaki Dry HLA Typing Trays are especially useful in areas where overnight shipping and freezer storage are problems.
  • Extensive coverage of Class I and Class II specificities
  • Robust reactions
  • Simple & Fast: one hour incubation
  • Requires only 1 µl of complement per well
  • Free lyophilized complement
  • Can be used for cell line typing
  • Durable under extended warmer shipping conditions
Terasaki Supplement HLA Tissue Typing Trays
The Supplement Trays work with other Terasaki HLA Typing Trays (T1, T2, and T3) to help solve difficult typing situations.
  • Contains antisera to verify rare and difficult antigens
  • Complete C locus typing tray
  • Accurate typing results
  • Superior technical support
  • Free prescreened rabbit complement
It all starts with a demo
Click here to request a demo for Terasaki HLA Tissue Typing Trays