One Lambda One Lambda™ Customer Portal
Search
clear search
Recent searches Clear History
Suggestions
All Results
Search
Learning Center Resources
Imlifidase for Kidney Transplantation of Highly Sensitized Patients With a Positive Crossmatch: The French Consensus Guidelines PubSummary
By: Dave Lowe, PhD, FRCPath 
Key Takeaways
  • Defines the criteria for harmonization of patient selection, associated treatments, and follow-up
  • Decision to use imlifidase is reliant upon the MFI values of the immunodominant donor-specific antibodies
  • Highlights the importance of dilution testing in ascertaining a more adequate reflection of antibody strength
Background
Imlifidase is a recombinant cysteine protease derived from Streptococcus pyogenes, which has the ability to cleave all classes and human IgG into F(ab’)2 and Fc fragments within 4–6 hours following administration. Additionally, imlifidase can act upon the IgG of the B cell receptor, and thus may abrogate the memory B cell response to antigenic stimulation. Consequently, imlifidase has recently received early access authorization in France to be used as a desensitization strategy for highly sensitized patients. This article summarizes the recent publication outlining the French guidelines to define the use of imlifidase, including patient selection criteria, monitoring frequencies, and associated treatment recommendations.
Recommendations
Eligible patients must have cPRA >98%, be under 65 years of age, and have been on the waitlist in excess of 3 years. The final decision to proceed with imlifidase treatment rests upon the MFI levels of the immunodominant donor-specific antibodies (DSAs).
Firstly, the immunodominant antibody must be >6,000 MFI; however, this must be reduced to a maximum of 5,000 after testing at 1:10 dilution. The guidelines recommend that this testing be performed using the One Lambda™ LABScreen™ single antigen assay. Secondly, the post-imlifidase complement-dependant cytotoxicity (CDC) crossmatch must be negative. Post-transplant, the guidelines recommend that antibody testing is performed frequently alongside surveillance biopsies in order to effectively monitor DSA rebound and the early detection of subclinical rejection.
Conclusion
Imlifidase has the potential to be a breakthrough treatment option for highly sensitized patients. However, more clinical data is required to be in a position to refine the protocols to optimize the use of this novel therapeutic. These French guidelines represent the start of the process needed to acquire this information.
What is the LABScreen Single Antigen assay?
Explore product
References
  1. Couzi L, Malvezzi P, Amrouche L, Anglicheau D, Blancho G, Caillard S, Freist M, Guidicelli GL, Kamar N, Lefaucheur C, Mariat C, Koenig A, Noble J, Thaunat O, Thierry A, Taupin JL, Bertrand D. Imlifidase for Kidney Transplantation of Highly Sensitized Patients With a Positive Crossmatch: The French Consensus Guidelines. Transpl Int. 2023 Jun 28;36:11244. doi: 10.3389/ti.2023.11244. PMID: 37448448; PMCID: PMC10336835.