Platindraht, 0.5 mm (0.02 Zoll) im Durchmesser, hart, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platindraht, 0.5 mm (0.02 Zoll) im Durchmesser, hart, Thermo Scientific Chemicals

Form: Ca. 0.44 g/10 cm, Härte: wie gezogen
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 cm
50 cm
250 cm
Katalognummer 010056.BS
auch als 010056-BS bezeichnet
Preis (EUR)
-
Menge:
10 cm
Großbestellung oder individuelle Größe anfordern
Specifications
AussehenSilvery white wire
CAS6-6-7440
Chemischer Name oder MaterialPlatinum
FarbeWeiß
MDL-NummerMFCD02091683
Mehr anzeigen
Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only