Thermo Scientific Chemicals

Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010188.18
50 g, Each
Thermo Scientific Chemicals

Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010188.18
50 g, Each
Quantity
Catalog number: 010188.18
also known as 010188-18
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7440-55-3
IUPAC Name
gallium
Molecular Formula
Ga
InChI Key
GYHNNYVSQQEPJS-UHFFFAOYSA-N
SMILES
[Ga]
Total Metal Impurities
0.01% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Substances/mixtures corrosive to metal
    Category 1
    Acute oral toxicity
    Category 4
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    H412 - Harmful to aquatic life with long lasting effects

    Physical hazards

    H290 - May be corrosive to metals

    Precautionary Statements

    P234 - Keep only in original packaging

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P273 - Avoid release to the environment

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P390 - Absorb spillage to prevent material damage

    P501 - Dispose of contents/ container to an approved waste disposal plant