Thermo Scientific Chemicals

Rhodium(III) 2,4-pentanedionate, Premion™, 99.99% (metals basis), Rh 25.2% min, Thermo Scientific Chemicals

Catalog number: 010561.06
5 g, Each
Thermo Scientific Chemicals

Rhodium(III) 2,4-pentanedionate, Premion™, 99.99% (metals basis), Rh 25.2% min, Thermo Scientific Chemicals

Catalog number: 010561.06
5 g, Each
Quantity
Catalog number: 010561.06
also known as 010561-06
Price (USD)

Chemical Identifiers

CAS
14284-92-5
IUPAC Name
rhodium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C15H21O6Rh
InChI Key
DGOINFUDFBWCMX-LNTINUHCSA-K
SMILES
[Rh+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Form
Crystalline
Total Metal Impurities
≤0.01%
Appearance (Color)
Yellow
Assay (special)
Rh 25.2-25.7%

Description

Rhodium(III) 2,4-pentanedionate is used in catalytic reagents for organic synthesis. Rhodium(III) 2,4-pentanedionate is useful for hydrosilylation of a variety of organic substrates like alkenes, terminal or internal acetylenes, conjugated dienes, alpha, beta -unsaturated aldehydes and allylic compounds. It performs the role of a catalyst in well-known reactions such as transformation processes including hydrogenation, ethanol steam reforming, CO oxidation, and NOx reduction. Rhodium acetylacetonate, as the organic solvent-soluble source of rhodium metal ions, can be used to prepare monodisperse size/shape controlled Rh nanocrystals. Rh nanocrystal monolayers have been reported to show high activity for hydrogenation of vinylcyclohexene to selectively produce ethylcyclohexene. The size/shape controlled nanocrystals find a variety of applications in oil refining processes, chemical synthesis and fuel cell technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text