Thermo Scientific Chemicals

Potassium bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010837.18
50 g, Each
Thermo Scientific Chemicals

Potassium bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010837.18
50 g, Each
Quantity
Catalog number: 010837.18
also known as 010837-18
Price (USD)
Price: 389.00
Online price: 330.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7758-02-3
IUPAC Name
potassium bromide
Molecular Formula
BrK
InChI Key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES
[K+].[Br-]
Total Metal Impurities
≤0.001% (10 ppm)
Assay (special)
?2.5 % of Theoretical value (K)
Appearance (Color)
White
Form
Crystals

Description

Potassium bromide is used in a variety of applications such as spectroscopy, infrared transmission, as a laboratory reagent, and for the removal of peripheral membrane proteins. It is also used in photosensitive chemicals, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text