Thermo Scientific Chemicals

Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010875.36
500 g, Each
Thermo Scientific Chemicals

Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010875.36
500 g, Each
Quantity
Catalog number: 010875.36
also known as 010875-36
Price (USD)
Price: 1,071.00
Online price: 909.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1330-43-4
IUPAC Name
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular Formula
B4Na2O7
InChI Key
NXSVMNYRVBQCMO-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Total Metal Impurities
0.002% max.

Description

Sodium tetraborate is used as a co-catalyst for (2,2,6,6-tetramethylpiperidin-1-yl)oxyl and sodium hypochlorite oxidation of alcohols in greener and non-chlorinated solvents. It acts as a reagent for the decomposition of oxides and silicates by fusion. It is used to prepare buffers, which is used in gel electrophoresis of DNA. It acts as a source of borate, micronutrient fertilizer, preservative in taxidermy, additive in ceramic slips and glazes. It is also used as a precursor for sodium perborate monohydrate and boric acid. It finds application as fluoride detoxification, neutron absorber in nuclear reactors and to make indelible ink for dip pens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text