Thermo Scientific Chemicals

Sodium metavanadate, typically 96%, V 38% min, Thermo Scientific Chemicals

Catalog number: 011092.30
250 g, Each
Thermo Scientific Chemicals

Sodium metavanadate, typically 96%, V 38% min, Thermo Scientific Chemicals

Catalog number: 011092.30
250 g, Each
Quantity
Catalog number: 011092.30
also known as 011092-30
Price (USD)
Price: 140.00
Online price: 119.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13718-26-8
IUPAC Name
sodium vanadiumoylolate
Molecular Formula
NaO3V
InChI Key
CMZUMMUJMWNLFH-UHFFFAOYSA-N
SMILES
[Na+].[O-][V](=O)=O
Assay (unspecified)
V 38% min

Description

Sodium metavanadate is used as an analytical reagent and mordant. It is also found to be useful in the photography industry and the pharmaceutical industry. In addition to this, it is used in the making of vanadium alloy and vanadium catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text