Cer(III)-Nitrathexahydrat, REacton, 99.5 % (REO), Thermo Scientific Chemicals
Cer(III)-Nitrathexahydrat, REacton, 99.5 % (REO), Thermo Scientific Chemicals
Cer(III)-Nitrathexahydrat, REacton, 99.5 % (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cer(III)-Nitrathexahydrat, REacton, 99.5 % (REO), Thermo Scientific Chemicals

Für die Trennung von Cer von anderen Seltenen Erden, Katalysator für die Hydrolyse von Phosphorsäureestern
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
100 g
500 g
1 kg
Katalognummer 011329.A1
auch als 011329-A1 bezeichnet
Preis (EUR)
329,65
Exklusiv online
366,00
Ersparnis 36,35 (10%)
Each
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
329,65
Exklusiv online
366,00
Ersparnis 36,35 (10%)
Each
Chemikalien-Kennzeichnungen
CAS10294-41-4
IUPAC Namecerium(3+) hexahydrate trinitrate
Molecular FormulaCeH12N3O15
InChI KeyQQZMWMKOWKGPQY-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥99.5% (CeO2/TREO)
Total Rare Earth Oxide Impurities≤0.5%
Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only