Calciumcarbonat,99.5 % (Metallbasis), Thermo Scientific Chemicals
Calciumcarbonat,99.5 % (Metallbasis), Thermo Scientific Chemicals
Calciumcarbonat,99.5 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calciumcarbonat,99.5 % (Metallbasis), Thermo Scientific Chemicals

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
2 kg
Katalognummer 011403.A3
auch als 011403-A3 bezeichnet
Preis (EUR)
470,00
Each
Menge:
2 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
470,00
Each
Chemikalien-Kennzeichnungen
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
FormPowder
Total Metal Impurities≤ 0.5%
Calcium carbonate is used in the purification of iron, refining of sugar from sugar beet and flue gas desulfurization applications in fossil fuel power stations. It is used in blackboard chalk, printing and writing paper, paints, plastics, PVC cables, polypropylene compounds, thermosetting resins, adhesives, sealants and decorating fillers. It is an abrasive and an ingredient in many glazes. It is also used in the pharmaceutical industry as an inert filler for tablets and other pharmaceuticals. It is used as a base in many organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only