Thermo Scientific Chemicals

Silver nitrite, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011418.09
10 g, Each
Thermo Scientific Chemicals

Silver nitrite, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011418.09
10 g, Each
Quantity
Catalog number: 011418.09
also known as 011418-09
Price (JPY)

Chemical Identifiers

CAS
7783-99-5
IUPAC Name
silver(1+) nitrite
Molecular Formula
AgNO2
InChI Key
KKKDGYXNGYJJRX-UHFFFAOYSA-M
SMILES
[Ag+].[O-]N=O
Total Metal Impurities
1% max.

Description

Silver nitrite is used in the silver plating process, hair dye, inks, permanent marker pens and determination of chloride ions in water and drilling fluids. It is used as Tollen’s reagent and used in the formation of aniline and nitro compounds in victor mayer reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text