Silbercarbonat, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Silbercarbonat, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Silbercarbonat, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Silbercarbonat, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silbercarbonat, 99.5 % (Metallbasis), Thermo Scientific Chemicals

CAS: 534-16-7 | CH2Ag2O3 | 277.76 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
Katalognummer 011420.14
auch als 011420-14 bezeichnet
Preis (EUR)
-
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS534-16-7
IUPAC Namecarbonic acid silver
Molecular FormulaCH2AgO3
InChI KeyLKZMBDSASOBTPN-UHFFFAOYSA-N
SMILES[Ag].OC(O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.5% max.
Silver carbonate is used in the production of silver powder in microelectronics, conducting polymers, and in tooth surface treatment to prevent cavities. It is also used as an antimicrobial agent for infection resistant surgical fabric materials. It is used in organic synthesis, and in biological specimen staining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only