Thermo Scientific Chemicals

Feuille d’argent, 0,025 mm (0,001 po) d’épaisseur, rigide, Premion™, 99,998 % (base de métaux), Thermo Scientific Chemicals

Référence: 011498.FF
25 x 25 mm, Each
Thermo Scientific Chemicals

Feuille d’argent, 0,025 mm (0,001 po) d’épaisseur, rigide, Premion™, 99,998 % (base de métaux), Thermo Scientific Chemicals

Référence: 011498.FF
25 x 25 mm, Each
Quantité
Référence: 011498.FF
également connu sous le numéro 011498-FF
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
7440-22-4
IUPAC Name
silver
Molecular Formula
Ag
InChI Key
BQCADISMDOOEFD-UHFFFAOYSA-N
SMILES
[Ag]
Total Metal Impurities
0.002% max.
Comment
0.025mm (0.001in) thick

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text