Citric acid monohydrate, 99.5-100.5% (by anhydrous basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Citric acid monohydrate, 99.5-100.5% (by anhydrous basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
2.5 kg
12 kg
50 kg
Katalognummer 0115-07
Preis (EUR)
-
Menge:
12 kg
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
Specifications
CAS5949-29-1
Chemischer Name oder MaterialCitric acid monohydrate
FarbeWeiß
Schmelzpunkt137°C to 152°C
SummenformelC6H8O7·H2O
VerpackungPolyethylene Pail with Screw Top
SynonymHydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
GüteMulti-Kompendium
Molekulargewicht (g/mol)210.14
Reinheit (%)99.5 to 100.5%
Menge12 kg
Unit SizeEach