Thermo Scientific Chemicals

Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011536.36
500 g, Each
Thermo Scientific Chemicals

Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011536.36
500 g, Each
Quantity
Catalog number: 011536.36
also known as 011536-36
Price (USD)
Price: 114.00
Online price: 96.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10025-69-1
IUPAC Name
λ²-tin(2+) dihydrate dichloride
Molecular Formula
Cl2H4O2Sn
InChI Key
FWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Sn++]
Assay (unspecified)
96% min

Description

Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text