Kaliumchlorid, ACS, 99.0 bis 100.5 %, Thermo Scientific Chemicals
Kaliumchlorid, ACS, 99.0 bis 100.5 %, Thermo Scientific Chemicals
Kaliumchlorid, ACS, 99.0 bis 100.5 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kaliumchlorid, ACS, 99.0 bis 100.5 %, Thermo Scientific Chemicals

In Pufferlösungen, Elektrodenzellen, der Fotografie, als Laborreagenz, in Düngemitteln, bei der Spektroskopie
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 g
1 kg
5 kg
Katalognummer 011595.A1
auch als 011595-A1 bezeichnet
Preis (EUR)
69,65
Exklusiv online
77,20
Ersparnis 7,55 (10%)
Each
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
69,65
Exklusiv online
77,20
Ersparnis 7,55 (10%)
Each
Chemikalien-Kennzeichnungen
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Impurity content(Barium, Ba) : Passes test (limit about 0.001%)
Impurity content(Iodide, I) : 0.002% max
Impurity content(Iron, Fe) : 3ppm max
Impurity content(Magnesium, Mg) : 0.001% max
Impurity content(Phosphate, PO₄) : 5ppm max
Mehr anzeigen
Potassium chloride is used to replenish electrolytes and restore water electrolyte balance in treating hypokalemia. It is also used in buffers, fertilizers and explosives and finds useful in medicine, scientific applications, and food processing. It plays an important role for the extraction and solubilization of proteins. It acts as a fire extinguishing agent in portable and wheeled fire extinguishers. It may be used as a softener unit in household water, which is an alternative to sodium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In Pufferlösungen, Elektrodenzellen, der Fotografie, als Laborreagenz, in Düngemitteln, bei der Spektroskopie
RUO – Research Use Only