Thermo Scientific Chemicals

Potassium iodate, ACS, 99.4-100.4%, Thermo Scientific Chemicals

Catalog number: 011602.36
500 g, Each
Thermo Scientific Chemicals

Potassium iodate, ACS, 99.4-100.4%, Thermo Scientific Chemicals

Catalog number: 011602.36
500 g, Each
Quantity
Catalog number: 011602.36
also known as 011602-36
Price (USD)
Price: 181.00
Online price: 153.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7758-05-6
IUPAC Name
potassium iodate
Molecular Formula
IKO3
InChI Key
JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES
[K+].[O-][I](=O)=O
Assay (unspecified)
99.4-100.4% KIO₃
Impurity content
Fe 0.001%
Impurity content
Nitrogen compounds (as N) 0.005%
Impurity content
Chloride and bromide (as Cl) 0.01%
Impurity content
I 0.001%

Description

Potassium iodate is an oxidizing agent used to prepare 2-styrylchromones from o-hydroxy-ω-cinnamylideneacetophenones by oxidative cyclization. It is used to iodinate table salt in order to prevent iodine deficiency. It is an active ingredient in baby formula milk. As a maturing agent, it is used in baking. Furthermore, it is involved in the protection of radioactive iodine accumulation in the thyroid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text