Magnesiumperchlorat, wasserfrei, ACS, Thermo Scientific Chemicals
Magnesiumperchlorat, wasserfrei, ACS, Thermo Scientific Chemicals
Magnesiumperchlorat, wasserfrei, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesiumperchlorat, wasserfrei, ACS, Thermo Scientific Chemicals

Magnesiumperchlorat, wasserfrei, Cl2MgO8, CAS-Nummer-10034-81-8, Magnesiumperchlorat, wasserfrei, wasserfreies Magnesiumperchlorat, Anhydron, Magnesiumperchlorat, Magnesiumdiperchlorat, HSDB 661, Dehydrite, perchlorat de magnesium französisch, Perchlorsäure, Magnesiumsalz, unii-7n77z541yf, 500 g
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
Katalognummer 011636.22
Preis (EUR)
90,20
Each
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
90,20
Each
Chemikalien-Kennzeichnungen
CAS10034-81-8
IUPAC Namemagnesium(2+) diperchlorate
Molecular FormulaCl2MgO8
InChI KeyMPCRDALPQLDDFX-UHFFFAOYSA-L
SMILES[Mg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
FormPowder, crystalline powder or granules or flakes and/or chunks
Titrable base≤0.025 meq/g
Titrable free acid≤0.005 meq/g
Loss on Drying≤8.0% (1g, 190±10°C)
Mehr anzeigen
Magnesium perchlorate is commonly utilized as a potent oxidizing agent. It is used as dehydrating agent in organic synthesis. It is practiced in many domains like pharma industry, pyrotechnics industry,in rechargeable batteries and it is a versatile tool in organic synthesis. It possesses photoelectronic and optical properties. Thus it can be used in photo electronic and optoelectronic devices. It is applied as a strong drying agent for gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only