Chromsilizid, > 99 % (Metallbasis), Thermo Scientific Chemicals
Chromsilizid, > 99 % (Metallbasis), Thermo Scientific Chemicals
Chromsilizid, > 99 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromsilizid, > 99 % (Metallbasis), Thermo Scientific Chemicals

CAS: 12018-09-6 | CrSi2 | 108.166 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
Katalognummer 012151.18
auch als 012151-18 bezeichnet
Preis (EUR)
69,10
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
69,10
Each
Chemikalien-Kennzeichnungen
CAS12018-09-6
Molecular FormulaCrSi2
InChI KeyCHXGWONBPAADHP-UHFFFAOYSA-N
SMILES[Si]#[Cr]#[Si]
Molecular Weight (g/mol)108.17
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities1% max.
Chromium silicide is applied in production of ceramic materials. It can also produce an oxidation-resistant surface in a low pressure plasma spraying.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only