Thermo Scientific Chemicals

Mercury(II) chloride, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 012274.36
500 g, Each
Thermo Scientific Chemicals

Mercury(II) chloride, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 012274.36
500 g, Each
Quantity
Catalog number: 012274.36
also known as 012274-36
Price (USD)

Chemical Identifiers

CAS
7487-94-7
IUPAC Name
mercury(2+) dichloride
Molecular Formula
Cl2Hg
InChI Key
LWJROJCJINYWOX-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Hg++]
Impurity content
Fe 0.002%
Assay (unspecified)
99.5% Cl₂Hg min
Comment
Residue after reduction 0.02%
Comment
Solution in ethyl ether P.T.

Description

Mercury(II) chloride is used as a catalyst in making plastics, depolarizer in batteries and as a reagent in organic synthesis and analytical chemistry. It is also used to prepare an amalgam with metals such as aluminum, which finds application as reducing agent in organic synthesis. It also finds application in photography as a photographic intensifier. Further, it serves as a preservative and used in the treatment of syphilis. It is also used to remove dithiane groups bonded to a carbonyl in an umpolung reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text