Thermo Scientific Chemicals

Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012283.36
500 g, Each
Thermo Scientific Chemicals

Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012283.36
500 g, Each
Quantity
Catalog number: 012283.36
also known as 012283-36
Price (USD)
Price: 154.00
Online price: 130.65
Your price:
Quantity
-

Chemical Identifiers

CAS
18282-10-5
IUPAC Name
stannanedione
Molecular Formula
O2Sn
InChI Key
XOLBLPGZBRYERU-UHFFFAOYSA-N
SMILES
O=[Sn]=O
Total Metal Impurities
0.1% max.

Description

It is employed in the manufacture of white enamels and for polishing glass. It has a broad range of applications in many areas such as semiconductors, photovoltaic materials, solar cells, lithium ion batteries, photocatalysts, and as gas-sensing agents in solid-state detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text