Thermo Scientific Chemicals

Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012524.A3
2 kg, Each
Thermo Scientific Chemicals

Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012524.A3
2 kg, Each
Quantity
Catalog number: 012524.A3
also known as 012524-A3
Price (USD)
Price: 854.00
Online price: 738.65
Your price:
Quantity
-

Chemical Identifiers

CAS
21041-93-0
IUPAC Name
λ²-cobalt(2+) dihydroxide
Molecular Formula
CoH2O2
InChI Key
ASKVAEGIVYSGNY-UHFFFAOYSA-L
SMILES
[OH-].[OH-].[Co++]
Total Metal Impurities
0.1% max.

Description

Cobalt(II) hydroxide is used as a drying agent for paints and varnishes. It acts as an aid to dry the lithographic inks and catalyst in the preparation of battery electrode impregnating solutions. It is also involved in the synthesis of other cobalt compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text