Thermo Scientific Chemicals

Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals

Catalog number: 012613.30
250 g, Each
Thermo Scientific Chemicals

Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals

Catalog number: 012613.30
250 g, Each
Quantity
Catalog number: 012613.30
also known as 012613-30
Price (USD)
Price: 315.00
Online price: 267.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13410-01-0
Total Metal Impurities
0.2% max.

Description

Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text