Thermo Scientific Chemicals

Siliciuro de cobre, piezas de color plata, 99,5 % (base metálica), Thermo Scientific Chemicals

Número de catálogo: 012844.22
100 g, Each
Thermo Scientific Chemicals

Siliciuro de cobre, piezas de color plata, 99,5 % (base metálica), Thermo Scientific Chemicals

Número de catálogo: 012844.22
100 g, Each
Cantidad
Número de catálogo: 012844.22
también denominado 012844-22
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
12159-07-8
IUPAC Name
pentacopper silicon
Molecular Formula
Cu5Si
InChI Key
JUZTWRXHHZRLED-UHFFFAOYSA-N
SMILES
[Si].[Cu].[Cu].[Cu].[Cu].[Cu]
Total Metal Impurities
0.5% max.

Descripción

Copper silicide thin film is used for passivation of copper-based chips, where it serves to suppress diffusion and electromigration and serves as a diffusion barrier. It is involved in the direct process, the industrial route to organosilicon compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention