Indiumarsenid, 99.9999 % (Metallbasis), Thermo Scientific Chemicals
Indiumarsenid, 99.9999 % (Metallbasis), Thermo Scientific Chemicals
Indiumarsenid, 99.9999 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indiumarsenid, 99.9999 % (Metallbasis), Thermo Scientific Chemicals

CAS: 1303-11-3 | AsIn | 189.74 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2 g
10 g
Katalognummer 012932.04
auch als 012932-04 bezeichnet
Preis (EUR)
145,00
Each
Menge:
2 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
145,00
Each
Chemikalien-Kennzeichnungen
CAS1303-11-3
IUPAC Nameindiganylidynearsane
Molecular FormulaAsIn
InChI KeyRPQDHPTXJYYUPQ-UHFFFAOYSA-N
SMILES[As]#[In]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.0001% max.
Indium arsenide is used for construction of infrared detectors, for the wavelength range of 1-3μm. It is widely used as terahertz radiation source as it is a strong photo-Dember emitter. Indium arsenide is also used for making of diode laser.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only