Kaliumperoxydisulfat, ACS, 99.0 % min., Thermo Scientific Chemicals
Kaliumperoxydisulfat, ACS, 99.0 % min., Thermo Scientific Chemicals
Kaliumperoxydisulfat, ACS, 99.0 % min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kaliumperoxydisulfat, ACS, 99.0 % min., Thermo Scientific Chemicals

Ein Polymerisationsbeschleuniger für PAGE-Gele.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 g
1 kg
Katalognummer 013145.A1
auch als 013145-A1 bezeichnet
Preis (EUR)
161,00
Each
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
161,00
Each
Chemikalien-Kennzeichnungen
CAS7727-21-1
IUPAC Namedipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaK2O8S2
InChI KeyUSHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)99.0% K₂S₂O₈ min
Trace Metals(as Pb) 0.001%
Impurity contentChlorine compounds (as Cl) 0.001%
Impurity contentFe 5ppm
Impurity contentMn 2ppm
Mehr anzeigen
A polymerization accelerant for PAGE gels. Potassium peroxydisulfate is used as an oxidizing agent in organic synthesis. It is involved in Elbs persulfate oxidation of phenols and the Boyland-Sims oxidation of anilines. Upon solution, it gives radicals and is used to initiate polymerization reactions to prepare styrene-butadiene rubber and polytetrafluoroethylene. It is used as bleaching agent in various hair bleaches, ion exchange agents, plating agents and surface treating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only