Thermo Scientific Chemicals

Gadolinium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 013208.22
100 g, Each
Thermo Scientific Chemicals

Gadolinium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 013208.22
100 g, Each
Quantity
Catalog number: 013208.22
also known as 013208-22
Price (USD)
Price: 265.00
Online price: 229.65
Your price:
Quantity
-

Chemical Identifiers

CAS
30010-20-9
IUPAC Name
gadolinium(3+) tribromide
Molecular Formula
Br3Gd
InChI Key
KGOKDPWKDBWITQ-UHFFFAOYSA-K
SMILES
[Br-].[Br-].[Br-].[Gd+3]
Total Rare Earth Oxide Impurities
0.01% max.

Description

It acts as dopant material. It is employed in gamma-ray spectroscopy, medical imaging, nuclear and high energy physics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text