Thermo Scientific Chemicals

Gallium(III) sulfide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 013387.02
0.5 g, Each
Thermo Scientific Chemicals

Gallium(III) sulfide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 013387.02
0.5 g, Each
Quantity
Catalog number: 013387.02
also known as 013387-02
Price (USD)
Quantity
-

Chemical Identifiers

CAS
12024-22-5
IUPAC Name
digallium(3+) trisulfanediide
Molecular Formula
Ga2S3
InChI Key
BVSHTEBQPBBCFT-UHFFFAOYSA-N
SMILES
[S--].[S--].[S--].[Ga+3].[Ga+3]
Total Metal Impurities
0.01% max.

Description

Gallium(III) sulfide is used in electronics and photonics. It acts as an active material in lithium secondary batteries. Further, it is used in thin-film solar cells. It is also used to prepare gallium lanthanum sulfide glass by reacting with lanthanum sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text