Lithium-Aluminiumoxid, weißes Pulver, Thermo Scientific Chemicals
Lithium-Aluminiumoxid, weißes Pulver, Thermo Scientific Chemicals
Lithium-Aluminiumoxid, weißes Pulver, Thermo Scientific Chemicals
Lithium-Aluminiumoxid, weißes Pulver, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium-Aluminiumoxid, weißes Pulver, Thermo Scientific Chemicals

CAS: 12003-67-7 | AlLiO2 | 65.92 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
1 kg
Katalognummer 013412.18
auch als 013412-18 bezeichnet
Preis (EUR)
106,00
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
106,00
Each
Chemikalien-Kennzeichnungen
CAS12003-67-7
IUPAC Name[(oxidoalumanylidene)oxo]lithiumylium
Molecular FormulaAlLiO2
InChI KeyYQNQTEBHHUSESQ-UHFFFAOYSA-N
SMILES[Li+].[O-][Al]=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)white
Formcrystalline solid
It is used as an inert electrolyte support material in molten carbonate fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only