Lithium aluminum hydride, 95% min
Lithium aluminum hydride, 95% min
Lithium aluminum hydride, 95% min
Lithium aluminum hydride, 95% min
Thermo Scientific Chemicals

Lithium aluminum hydride, 95% min

Reducing Agent For Pharmaceuticals, Perfumes, and Fine organic Chemicals; Source of Hydrogen | CAS: 16853-85-3 | AlH4Li | 37.95 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
500 g
5 x 25 g
5 x 100 g
Katalognummer 013442.36
auch als 013442-36 bezeichnet
Preis (EUR)
-
Menge:
500 g
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS16853-85-3
IUPAC Namelithium(1+) alumanuide
Molecular FormulaAlH4Li
InChI KeyOCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES[Li+].[AlH4-]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)95% min
Reducing agent for pharmaceuticals, perfumes and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizationsLithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It is also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium, which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only