Thermo Scientific Chemicals

Sodium metaperiodate, 98%, Thermo Scientific Chemicals

Catalog number: 013798.A3
2 kg, Each
Thermo Scientific Chemicals

Sodium metaperiodate, 98%, Thermo Scientific Chemicals

Catalog number: 013798.A3
2 kg, Each
Quantity
Catalog number: 013798.A3
Price (USD)
Price: 750.00
Online price: 637.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7790-28-5
IUPAC Name
sodium periodate
Molecular Formula
INaO4
InChI Key
JQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES
[Na+].[O-][I](=O)(=O)=O
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder
Assay (special)
≥97.5%

Description

Source of periodic acid, as an analytical reagent, and as an oxidizing agent Sodium metaperiodate is used as an oxidizing reagent involved in the oxidation of cellulose. It is involved in the cleavage of vicinal diols to prepare two aldehydes. This oxidation is commonly used in selectively label RNA, saccharides and other compounds with vicinal diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text