Thermo Scientific Chemicals

Nickel Chromium wire, 0.51mm (0.02in) dia, Thermo Scientific Chemicals

Catalog number: 013967.H9
500 m, Each
Thermo Scientific Chemicals

Nickel Chromium wire, 0.51mm (0.02in) dia, Thermo Scientific Chemicals

Catalog number: 013967.H9
500 m, Each
Quantity
Catalog number: 013967.H9
also known as 013967-H9
Price (USD)
Price: 376.00
Online price: 319.65
Your price:
Quantity
-

Chemical Identifiers

CAS
11106-97-1
IUPAC Name
chromium nickel
Molecular Formula
CrNi
InChI Key
VNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES
[Cr].[Ni]
Form
Wire, 0.51 mm diameter
Assay from Suppliers CofA
Chromium: 17.5-22.5%
Assay from Suppliers CofA
Nickel: 77.5-82.5%

Description

These alloys are typically used to increase the resistivity of wire. They are also used in some dental restorations (fillings). It is commonly used in ceramic as an internal support. Nichrome wire is used for its ability to withstand the high temperatures that occur when clay work is fired in a kiln.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text