Kobalt(IV)-sulfid, 99.5 % (Metallbasis ohne Ni), Ni < 0.2 %, Thermo Scientific Chemicals
Kobalt(IV)-sulfid, 99.5 % (Metallbasis ohne Ni), Ni < 0.2 %, Thermo Scientific Chemicals
Kobalt(IV)-sulfid, 99.5 % (Metallbasis ohne Ni), Ni < 0.2 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kobalt(IV)-sulfid, 99.5 % (Metallbasis ohne Ni), Ni < 0.2 %, Thermo Scientific Chemicals

Have Questions?
KatalognummerMenge
014005.06
auch als 014005-06 bezeichnet
5 g
Katalognummer 014005.06
auch als 014005-06 bezeichnet
Preis (EUR)
141,00
Each
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
141,00
Each
Chemikalien-Kennzeichnungen
CAS12013-10-4
IUPAC Namedisulfanylidenecobalt
Molecular FormulaCoS2
InChI KeyXUKVMZJGMBEQDE-UHFFFAOYSA-N
SMILESS=[Co]=S
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Black
Total Metal Impurities≤0.5%
Particle Size-200 mesh (≤74 microns) at D90 (Diameter at 90%)
FormPowder
Cobalt(IV) sulfide is a promising battery electrode material exhibiting good electrochemical properties for lithium sulfur batteries, thermal batteries and lithium ion secondary battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only